Recombinat Protein,Mouse gp130 CD130 IL-6 R beta Protein, His Tag,Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Glu 617 (Accession NP034690).This protein carries a polyhistidine tag at the C-te 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse gp130 CD130 IL-6 R beta Protein, His Tag,Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Glu 617 (Accession NP034690).This protein carries a polyhistidine tag at the C-te 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse gp130 CD130 IL-6 R beta Protein, His Tag,Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Glu 617 (Accession NP034690).This protein carries a polyhistidine tag at the C-te 1 * 100 µG ACRBILT-M52H1100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou