Human ITGAVITGB1 Heterodimer Protein, His TagTag Free (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human ITGAVITGB1 Heterodimer Protein, His TagTag Free (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human ITGAVITGB1 Heterodimer Protein, His TagTag Free (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 100 µG ACRBIT1-H52E1100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou