Human ITGA10ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Ser 1122 (ITGA10) Gln 21 - Asp 728 (ITGB1) (Accession O75578-1 (ITGA10) P05556-1 (ITGB1)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ITGA10ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Ser 1122 (ITGA10) Gln 21 - Asp 728 (ITGB1) (Accession O75578-1 (ITGA10) P05556-1 (ITGB1)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ITGA10ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Ser 1122 (ITGA10) Gln 21 - Asp 728 (ITGB1) (Accession O75578-1 (ITGA10) P05556-1 (ITGB1)). 1 * 1 mg ACRBIT1-H52WA-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou