Human ITGAVITGB6 Heterodimer Protein, His TagTag Free (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 22 - Asn 707 (ITGB6) (Accession NP002201.1 (ITGAV) AAI21179.1 (ITGB6)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ITGAVITGB6 Heterodimer Protein, His TagTag Free (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 22 - Asn 707 (ITGB6) (Accession NP002201.1 (ITGAV) AAI21179.1 (ITGB6)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ITGAVITGB6 Heterodimer Protein, His TagTag Free (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 22 - Asn 707 (ITGB6) (Accession NP002201.1 (ITGAV) AAI21179.1 (ITGB6)). 1 * 1 mg ACRBIT6-H52E1-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou