Human ITGAEITGB7 Heterodimer Protein, His TagTag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) Glu 20 - His 723 (ITGB7) (Accession P38570-1 (ITGAE) P26010-1 (ITGB7)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ITGAEITGB7 Heterodimer Protein, His TagTag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) Glu 20 - His 723 (ITGB7) (Accession P38570-1 (ITGAE) P26010-1 (ITGB7)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ITGAEITGB7 Heterodimer Protein, His TagTag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) Glu 20 - His 723 (ITGB7) (Accession P38570-1 (ITGAE) P26010-1 (ITGB7)). 1 * 1 mg ACRBIT7-H52W7-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou