Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 1 mg ACRBIT8-M52W6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou