Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK2 93). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK2 93). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg ACRBJA1-H52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou