Human VEGF R2, His Tag (HPLC-verified) (KDR-H5227) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Acce ssion AAI31823.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human VEGF R2, His Tag (HPLC-verified) (KDR-H5227) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Acce ssion AAI31823.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human VEGF R2, His Tag (HPLC-verified) (KDR-H5227) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Acce ssion AAI31823.1). 1 * 100 µG ACRBKDR-H5227100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou