Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhis 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhis 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag™,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhis 1 * 25 µG ACRBLA1-H82E3-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou