Human LAG-3, His Tag (LA3-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession P18627-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human LAG-3, His Tag (LA3-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession P18627-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human LAG-3, His Tag (LA3-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession P18627-1). 1 * 100 µG ACRBLA3-H5222100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou