Recombinat Protein,Mouse LAG-3 CD223 Protein, His Tag,Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession Q61790).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse LAG-3 CD223 Protein, His Tag,Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession Q61790).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse LAG-3 CD223 Protein, His Tag,Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession Q61790).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg ACRBLA3-M52H5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou