Recombinat Protein,Rat LAG3 CD223 Protein, His Tag,Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It con tains AA Gly 24 - Leu 442 (Accession NP997678).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rat LAG3 CD223 Protein, His Tag,Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It con tains AA Gly 24 - Leu 442 (Accession NP997678).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rat LAG3 CD223 Protein, His Tag,Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It con tains AA Gly 24 - Leu 442 (Accession NP997678).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 100 µG ACRBLA3-R52H5100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou