Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hu man 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag a 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hu man 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag a 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hu man 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag a 1 * 100 µG ACRBLBP-H52H5100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou