Recombinat Protein,Human Lipocalin-2 NGAL Protein, His Tag,Human Lipocalin-2, His Tag (LC2-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 198 (Accession NP005555.2).This protein carries a polyhistidine tag at the C-t r 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Lipocalin-2 NGAL Protein, His Tag,Human Lipocalin-2, His Tag (LC2-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 198 (Accession NP005555.2).This protein carries a polyhistidine tag at the C-t r 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Lipocalin-2 NGAL Protein, His Tag,Human Lipocalin-2, His Tag (LC2-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 198 (Accession NP005555.2).This protein carries a polyhistidine tag at the C-t r 1 * 1 mg ACRBLC2-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou