Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession XP0055 88053.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession XP0055 88053.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession XP0055 88053.1). 1 * 1 mg ACRBLDR-C5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou