Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession NP000518.1) . 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession NP000518.1) . 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession NP000518.1) . 1 * 1 mg ACRBLDR-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou