Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 100 µG ACRBLDR-M52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou