Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). 1 * 1 mg ACRBLE3-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou