Recombinat Protein,Human LILRB2 CD85d ILT4 Protein, His Tag,Human LILRB2, His Tag (LI2-H5220) is expressed from human 293 cells (H EK293). It contains AA Gln 22 - Val 461 (Accession AAH36827).This protein carries a polyhistidine tag at the C-terminu 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human LILRB2 CD85d ILT4 Protein, His Tag,Human LILRB2, His Tag (LI2-H5220) is expressed from human 293 cells (H EK293). It contains AA Gln 22 - Val 461 (Accession AAH36827).This protein carries a polyhistidine tag at the C-terminu 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human LILRB2 CD85d ILT4 Protein, His Tag,Human LILRB2, His Tag (LI2-H5220) is expressed from human 293 cells (H EK293). It contains AA Gln 22 - Val 461 (Accession AAH36827).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG ACRBLI2-H5220100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou