Recombinat Protein,Biotinylated Human LILRA3 CD85e Protein, His,Avitag,Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is expres sed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession AAH28208.1).This protein carries a po g 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human LILRA3 CD85e Protein, His,Avitag,Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is expres sed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession AAH28208.1).This protein carries a po g 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human LILRA3 CD85e Protein, His,Avitag™,Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is expres sed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession AAH28208.1).This protein carries a po g 1 * 25 µG ACRBLI3-H82E0-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou