Recombinat Protein,Mouse LIF Protein, His Tag,Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession P09056).This protein carries a polyhistidine tag at the C-terminus. The protein has o 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse LIF Protein, His Tag,Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession P09056).This protein carries a polyhistidine tag at the C-terminus. The protein has o 1 * 50 µG ACRBLIF-M5227-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou