Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag™,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 200 µG ACRBLO2-H82E3200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou