Recombinat Protein,Human LRRC4 Protein, His Tag,Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 39 - Lys 527 (Accession AAI11562.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human LRRC4 Protein, His Tag,Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 39 - Lys 527 (Accession AAI11562.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human LRRC4 Protein, His Tag,Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 39 - Lys 527 (Accession AAI11562.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg ACRBLR4-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou