Recombinat Protein,Human MBL Protein, His Tag,Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession AAH96179).This protein carries a polyhistidine tag at the C-terminus. The protein h M 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human MBL Protein, His Tag,Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession AAH96179).This protein carries a polyhistidine tag at the C-terminus. The protein h M 1 * 1 mg ACRBMBL-H5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou