Recombinat Protein,Human MERTK Mer Protein, His Tag,Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession Q12866-1).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human MERTK Mer Protein, His Tag,Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession Q12866-1).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human MERTK Mer Protein, His Tag,Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession Q12866-1).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg ACRBMEK-H52H6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou