Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 µG ACRBMIA-H5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou