Recombinat Protein,Biotinylated Human MIF Protein, His,Avitag,Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from huma n 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession P14174-1).This protein carries a polyhistidine tag a 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human MIF Protein, His,Avitag,Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from huma n 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession P14174-1).This protein carries a polyhistidine tag a 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human MIF Protein, His,Avitag™,Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from huma n 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession P14174-1).This protein carries a polyhistidine tag a 1 * 25 µG ACRBMIF-H82E7-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou