Recombinat Protein,Human Neprilysin MME CD10 Protein, Tag Free,Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).Human Neprilysin, with Gly-Pro at the N- terminu 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Neprilysin MME CD10 Protein, Tag Free,Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).Human Neprilysin, with Gly-Pro at the N- terminu 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Neprilysin MME CD10 Protein, Tag Free,Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).Human Neprilysin, with Gly-Pro at the N- terminu 1 * 1 mg ACRBMME-H521B-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou