Recombinat Protein,Human Neprilysin MME CD10 Protein, Fc Tag,Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cell s (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).This protein carries a human IgG1 Fc tag at the N-t 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Neprilysin MME CD10 Protein, Fc Tag,Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cell s (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).This protein carries a human IgG1 Fc tag at the N-t 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Neprilysin MME CD10 Protein, Fc Tag,Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cell s (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).This protein carries a human IgG1 Fc tag at the N-t 1 * 50 µG ACRBMME-H526A-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou