Recombinat Protein,Human Mesothelin MSLN (37-286) Protein, His Tag,Human Mesothelin (37-286), His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession AAH09272).This protein carries a polyhistidine er 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Mesothelin MSLN (37-286) Protein, His Tag,Human Mesothelin (37-286), His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession AAH09272).This protein carries a polyhistidine er 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Mesothelin MSLN (37-286) Protein, His Tag,Human Mesothelin (37-286), His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession AAH09272).This protein carries a polyhistidine er 1 * 1 mg ACRBMSN-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou