FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H522 1 * 25 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H522 1 * 25 µG ACRBMSN-HF223-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou