RFPFusion Human Mesothelin (296-580), His Tag (MSN-HR2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296-Gly 58 0 (Accession AAH09272.1). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
RFPFusion Human Mesothelin (296-580), His Tag (MSN-HR2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296-Gly 58 0 (Accession AAH09272.1). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
RFP Fusion Human Mesothelin (296-580), His Tag (MSN-HR2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296-Gly 58 0 (Accession AAH09272.1). 1 * 200 µG ACRBMSN-HR2P4200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou