Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 µG ACRBMU1-H5252100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou