Recombinat Protein,Human NKp46 NCR1 CD335 Protein, His Tag,Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK 293). It contains AA Gln 22 - Asn 254 (Accession AAH64806.1).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, His Tag,Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK 293). It contains AA Gln 22 - Asn 254 (Accession AAH64806.1).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, His Tag,Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK 293). It contains AA Gln 22 - Asn 254 (Accession AAH64806.1).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg ACRBNC1-H52H4-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou