Recombinat Protein,Biotinylated Human NKp46 NCR1 CD335 Protein, Fc,Avitag,Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expr essed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This protein carries a human 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human NKp46 NCR1 CD335 Protein, Fc,Avitag,Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expr essed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This protein carries a human 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human NKp46 NCR1 CD335 Protein, Fc,Avitag™,Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expr essed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This protein carries a human 1 * 200 µG ACRBNC1-H82F9200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou