Recombinat Protein,Human NKp30 NCR3 CD337 Protein, His Tag,Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK 293). It contains AA Leu 19 - Gly 135 (Accession AAH52582).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human NKp30 NCR3 CD337 Protein, His Tag,Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK 293). It contains AA Leu 19 - Gly 135 (Accession AAH52582).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NKp30 NCR3 CD337 Protein, His Tag,Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK 293). It contains AA Leu 19 - Gly 135 (Accession AAH52582).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg ACRBNC3-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou