Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C 1 * 1 mg ACRBNE4-C52H4-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou