Recombinat Protein,Human IL-12A NKSF1 p35 Protein, Fc Tag,Human IL-12A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK2 93). It contains AA Arg 23 - Ser 219 (Accession P29459).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IL-12A NKSF1 p35 Protein, Fc Tag,Human IL-12A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK2 93). It contains AA Arg 23 - Ser 219 (Accession P29459).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-12A NKSF1 p35 Protein, Fc Tag,Human IL-12A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK2 93). It contains AA Arg 23 - Ser 219 (Accession P29459).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 500 µG ACRBNK1-H5252500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou