Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg ACRBNKA-H5244-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou