Recombinat Protein,Human NKG2D CD314 Protein, His Tag,Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession P26718-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NKG2D CD314 Protein, His Tag,Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession P26718-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 100 µG ACRBNKD-H5245100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou