Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 1 mg ACRBNM1-H5147-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou