Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It con tains AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein t 1 * 100 µG ACRBNM1-H5147100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou