Recombinat Protein,Human TrkB NTRK2 Protein, His Tag,Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human TrkB NTRK2 Protein, His Tag,Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TrkB NTRK2 Protein, His Tag,Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 1 mg ACRBNT2-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou