Recombinat Protein,Biotinylated Human NTB-A SLAMF6 Protein, His,Avitag,Biotinylated Human NTB-A, His,Avitag (NTA-H82E6) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession Q96DU3-1).This protein carries a polyhis 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human NTB-A SLAMF6 Protein, His,Avitag,Biotinylated Human NTB-A, His,Avitag (NTA-H82E6) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession Q96DU3-1).This protein carries a polyhis 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human NTB-A SLAMF6 Protein, His,Avitag™,Biotinylated Human NTB-A, His,Avitag (NTA-H82E6) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession Q96DU3-1).This protein carries a polyhis 1 * 200 µG ACRBNTA-H82E6200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou