Recombinat Protein,Human Osteopontin (OPN) Protein, His Tag,Human Osteopontin, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession AAH07016).This protein carries a polyhistidine tag at the C-te t 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Osteopontin (OPN) Protein, His Tag,Human Osteopontin, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession AAH07016).This protein carries a polyhistidine tag at the C-te t 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Osteopontin (OPN) Protein, His Tag,Human Osteopontin, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession AAH07016).This protein carries a polyhistidine tag at the C-te t 1 * 100 µG ACRBOPN-H5227100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou