Human OX40, His Tag (OX0-H5224) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession P43489-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human OX40, His Tag (OX0-H5224) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession P43489-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human OX40, His Tag (OX0-H5224) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession P43489-1). 1 * 1 mg ACRBOX0-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou