Recombinat Protein,Biotinylated Human CD200 OX-2 Protein, Fc,Avitag,Biotinylated Human CD200, Fc,Avitag (OX2-H82F1) is expressed f rom human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession P41217-1).This protein carries a human IgG1 C 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human CD200 OX-2 Protein, Fc,Avitag,Biotinylated Human CD200, Fc,Avitag (OX2-H82F1) is expressed f rom human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession P41217-1).This protein carries a human IgG1 C 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human CD200 OX-2 Protein, Fc,Avitag™,Biotinylated Human CD200, Fc,Avitag (OX2-H82F1) is expressed f rom human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession P41217-1).This protein carries a human IgG1 C 1 * 25 µG ACRBOX2-H82F1-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou