Human OX40 Ligand, His Tag (active trimer) (MALS verified) (OXL-H52Q8) is expressed from human 293 cells (HEK293). It contains AA Gl n 51 - Leu 183 (Accession NP003317). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human OX40 Ligand, His Tag (active trimer) (MALS verified) (OXL-H52Q8) is expressed from human 293 cells (HEK293). It contains AA Gl n 51 - Leu 183 (Accession NP003317). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human OX40 Ligand, His Tag (active trimer) (MALS verified) (OXL-H52Q8) is expressed from human 293 cells (HEK293). It contains AA Gl n 51 - Leu 183 (Accession NP003317). 1 * 1 mg ACRBOXL-H52Q8-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou