Human PCSK9, His Tag (PC9-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human PCSK9, His Tag (PC9-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human PCSK9, His Tag (PC9-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). 1 * 1 mg ACRBPC9-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou