Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession AAH38085.1) . 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession AAH38085.1) . 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession AAH38085.1) . 1 * 1 mg ACRBPC9-M5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou